Dimethylphenylvinylsilane ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₀H₁₄Si Punto di ebollizione: 82°C Flash Pt: 59°C |
MDL Number:
MFCD00053857 CAS Number: 1125-26-4 UN: 1993 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...