Stampa…

2,4-Dimethoxy-5-pyrimidineboronic acid ≥95.0% (by titrimetric analysis) (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2,4-Dimethoxypyrimidine-5-boronic acid, (2,4-Dimethoxypyrimidin-5-yl)boronic acid, 2,4-Dimethoxy-5-pyrimidylboronic acid

D5632-1G D5632-5G
TCIAD5632-1GEA 34 EUR
TCIAD5632-1G TCIAD5632-5G
2,4-Dimethoxy-5-pyrimidineboronic acid ≥95.0% (by titrimetric analysis) (contains varying amounts of Anhydride)
2,4-Dimethoxy-5-pyrimidineboronic acid
Formula: C₆H₉BN₂O₄
Peso molecolare: 183,9577 g/mol
Punto di fusione: 117°C
MDL Number: MFCD01318985
CAS Number: 89641-18-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR