Stampa…

Disuccinimidyl Succinate ≥97.0% (by total nitrogen basis)

Fornitore: TCI EUROPE

Synonyms: Bis(2,5-dioxopyrrolidin-1-yl) succinate, Succinic acid Disuccinimidyl ester

D5622-1G D5622-250MG
TCIAD5622-1GEA 240 EUR
TCIAD5622-1G TCIAD5622-250MG
Disuccinimidyl Succinate ≥97.0% (by total nitrogen basis)
Disuccinimidyl Succinate
Formula: C₁₂H₁₂N₂O₈
Peso molecolare: 312,23 g/mol
Punto di fusione: 306°C
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00228233
CAS Number: 30364-60-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (with Total Nitrogen) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR