N,N-Dimethyl-1,3-propanediamine dihydrobromide ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
N1,N1-Dimethylpropane-1,3-diamine dihydrobromide, N,N-Dimethyl-1,3-propanediammonium Dibromide, 3-(Dimethylamino)propylamine dihydrobromide
D5618-1G
D5618-5G
TCIAD5618-1GEA
23
EUR
TCIAD5618-1G
TCIAD5618-5G
N,N-Dimethyl-1,3-propanediamine dihydrobromide ≥98.0% (by titrimetric analysis)
N,N-Dimethyl-1,3-propanediamine dihydrobromide
Formula:
C₅H₁₄N₂·₂HBr Peso molecolare: 264,01 g/mol |
CAS Number:
2710685-13-3 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...