Stampa…

N,N-Dimethylethylenediamine dihydrobromide ≥98.0% (by total nitrogen basis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: N,N-Dimethylethylenediammonium Dibromide, N1,N1-Dimethylethane-1,2-diamine dihydrobromide, 2-(Dimethylamino)ethylamine dihydrobromide

D5615-1G D5615-5G
TCIAD5615-1GEA 24 EUR
TCIAD5615-1G TCIAD5615-5G
N,N-Dimethylethylenediamine dihydrobromide ≥98.0% (by total nitrogen basis)
N,N-Dimethylethylenediamine dihydrobromide
Formula: C₄H₁₂N₂·₂HBr
Peso molecolare: 249,98 g/mol
CAS Number: 1245570-04-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR