N,N-Dimethyl-4-(trifluoromethyl)aniline ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
D5607-1G
D5607-5G
TCIAD5607-1GEA
110
EUR
TCIAD5607-1G
TCIAD5607-5G
N,N-Dimethyl-4-(trifluoromethyl)aniline ≥98.0% (by GC, titration analysis)
N,N-Dimethyl-4-(trifluoromethyl)aniline
Formula:
C₉H₁₀F₃N Peso molecolare: 189,18 g/mol |
MDL Number:
MFCD22053163 CAS Number: 329-17-9 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 69.0 to 73.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...