Stampa…

N,N-Dimethyl-2-(trifluoromethyl)aniline ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE

D5605-1G D5605-5G
TCIAD5605-1GEA 113 EUR
TCIAD5605-1G TCIAD5605-5G
N,N-Dimethyl-2-(trifluoromethyl)aniline ≥98.0% (by GC, titration analysis)
N,N-Dimethyl-2-(trifluoromethyl)aniline
Formula: C₉H₁₀F₃N
Peso molecolare: 189,18 g/mol
Punto di ebollizione: 170°C
MDL Number: MFCD01862096
CAS Number: 54672-14-9

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR