Stampa…

3,3'-Di(9H-carbazol-9-yl)-1,1'-biphenyl ≥99.0% (by HPLC, total nitrogen), purified by sublimation

Fornitore: TCI EUROPE
Pericolo

Synonyms: mCBP

D5594-1G D5594-200MG
TCIAD5594-1GEA 340 EUR
TCIAD5594-1G TCIAD5594-200MG
3,3'-Di(9H-carbazol-9-yl)-1,1'-biphenyl ≥99.0% (by HPLC, total nitrogen), purified by sublimation
3,3'-Di(9H-carbazol-9-yl)-1,1'-biphenyl
Formula: C₃₆H₂₄N₂
Peso molecolare: 484,60 g/mol
MDL Number: MFCD28138084
CAS Number: 342638-54-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (with Total Nitrogen) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR