Stampa…

(4R)-2,2-Dimethyl-4-thiazolidinecarboxylic acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

D5586-1G D5586-5G
TCIAD5586-1GEA 40 EUR
TCIAD5586-1G TCIAD5586-5G
(4R)-2,2-Dimethyl-4-thiazolidinecarboxylic acid ≥98.0% (by titrimetric analysis)
(4R)-2,2-Dimethyl-4-thiazolidinecarboxylic acid
Formula: C₆H₁₁NO₂S
Peso molecolare: 161,22 g/mol
CAS Number: 58166-38-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -164.0 to -171.0 deg (C=0.1, acetone)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR