Stampa…

(3,3-Difluorocyclobutyl)methanol ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

D5582-200MG D5582-1G
TCIAD5582-200MGEA 49 EUR
TCIAD5582-200MG TCIAD5582-1G
(3,3-Difluorocyclobutyl)methanol ≥98.0% (by GC)
(3,3-Difluorocyclobutyl)methanol
Formula: C₅H₈F₂O
MDL Number: MFCD11877861
CAS Number: 681128-39-2
UN: 1987
ADR: 3,III

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR