Stampa…

Dinotefuran ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-Methyl-3-nitro-2-(tetrahydro-3-furanylmethyl)guanidine

D5560-1G D5560-5G
TCIAD5560-1GEA 87 EUR
TCIAD5560-1G TCIAD5560-5G
Dinotefuran ≥98.0% (by HPLC, titration analysis)
Dinotefuran
Formula: C₇H₁₄N₄O₃
Peso molecolare: 202,2 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD06795001
CAS Number: 165252-70-0
UN: 3077
ADR: 9,III
Merck Index: 14,03289

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 104.0 to 108.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR