Stampa…

3-[(3R,4R)-3,4-Dimethylpiperidin-4-yl]phenol ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (3R,4R)-3,4-Dimethyl-4-(3-hydroxyphenyl)piperidine

D5558-200MG D5558-1G
TCIAD5558-200MGEA 93 EUR
TCIAD5558-200MG TCIAD5558-1G
3-[(3R,4R)-3,4-Dimethylpiperidin-4-yl]phenol ≥98.0% (by GC)
3-[(3R,4R)-3,4-Dimethylpiperidin-4-yl]phenol
Formula: C₁₃H₁₉NO
Peso molecolare: 205,30 g/mol
MDL Number: MFCD06659960
CAS Number: 119193-19-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 181.0 to 185.0 deg-C
Specific rotation [a]20/D +114 to +125 deg (C=1, methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR