BDP FL ≥97.0% (by HPLC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4,4-Difluoro-5,7-dimethyl-4-bora-3a,4a-diaza-s-indacene-3-propionic acid
D5554-5MG
D5554-25MG
TCIAD5554-5MGEA
228
EUR
TCIAD5554-5MG
TCIAD5554-25MG
BDP FL ≥97.0% (by HPLC)
BDP FL
Formula:
C₁₄H₁₅BF₂N₂O₂ Peso molecolare: 292,09 g/mol Punto di fusione: 196°C |
MDL Number:
MFCD00467384 CAS Number: 165599-63-3 |
Risultati del test di specificazione
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Elemental analysis (Nitrogen) | 9.10 to 10.10 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...