Stampa…

3,6-Dibromo-9-(2-naphthalenyl)-9H-carbazole ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3,6-Dibromo-9-(naphthalen-2-yl)-9H-carbazole

D5546-1G D5546-5G
TCIAD5546-1GEA 57 EUR
TCIAD5546-1G TCIAD5546-5G
3,6-Dibromo-9-(2-naphthalenyl)-9H-carbazole ≥98.0% (by HPLC, total nitrogen)
3,6-Dibromo-9-(2-naphthalenyl)-9H-carbazole
Formula: C₂₂H₁₃Br₂N
Peso molecolare: 451,16 g/mol
MDL Number: MFCD29072769
CAS Number: 1221237-83-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 197.0 to 201.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR