4,4-Difluorocyclohexanol ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
D5540-200MG
D5540-1G
TCIAD5540-200MGEA
39
EUR
TCIAD5540-200MG
TCIAD5540-1G
4,4-Difluorocyclohexanol ≥98.0% (by GC)
4,4-Difluorocyclohexanol
Formula:
C₆H₁₀F₂O Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD10000566 CAS Number: 22419-35-8 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to lump |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...