2,7-Dibromodibenzo[b,e][1,4]dioxin ≥98.0% (by GC)
Fornitore: TCI
Formula:
C₁₂H₆Br₂O₂ |
CAS Number:
39073-07-9 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 194.0 to 198.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...