Stampa…

Doxercalciferol ≥98.0% (by HPLC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: (1R,3S,Z)-5-((E)-2-((1R,3aS,7aR)-1-((2R,5R,E)-5,6-Dimethylhept-3-en-2-yl)-7a-methylhexahydro-1H-inden-4(2H)-ylidene)ethylidene)-4-methylenecyclohexane-1,3-diol, (5Z,7E,22E)-9,10-Secoergosta-5,7,10,22-tetraene-1,3-diol

D5526-5MG D5526-25MG
TCIAD5526-5MGEA 147 EUR
TCIAD5526-5MG TCIAD5526-25MG
Doxercalciferol ≥98.0% (by HPLC)
Doxercalciferol
Formula: C₂₈H₄₄O₂
Peso molecolare: 412,66 g/mol
Punto di fusione: 140°C
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00871065
CAS Number: 54573-75-0
UN: 2811
ADR: 6.1,I
Merck Index: 14,03436

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D +46.0 to +50.0 deg (C=0.5,EtOH)
Elemental analysis (Carbon) 79.90 to 83.10 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR