Stampa…

3-(3,5-Di-tert-butyl-4-hydroxyphenyl)-N'-[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoyl]propanehydrazide ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: Antioxidant 1024, 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)-N'-(3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoyl)propanehydrazide, 1,2-Bis(3,5-di-tert-butyl-4-hydroxyhydrocinnamoyl)hydrazine

D5516-5G D5516-25G
TCIAD5516-5GEA 34 EUR
TCIAD5516-5G TCIAD5516-25G
3-(3,5-Di-tert-butyl-4-hydroxyphenyl)-N'-[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoyl]propanehydrazide ≥98.0% (by HPLC)
3-(3,5-Di-tert-butyl-4-hydroxyphenyl)-N'-[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoyl]propanehydrazide
Formula: C₃₄H₅₂N₂O₄
Peso molecolare: 552,80 g/mol
MDL Number: MFCD00072662
CAS Number: 32687-78-8
EINECS: 251-156-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 226.0 to 230.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR