3,4-Dibromoiodobenzene ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₆H₃Br₂I |
MDL Number:
MFCD09753707 CAS Number: 909072-74-8 |
Risultati del test di specificazione
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 58.0 to 62.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...