Stampa…

Dihydrorhodamine 6G

Fornitore: TCI EUROPE

Synonyms: 2-[3,6-Bis(ethylamino)-2,7-dimethyl-9H-xanthen-9-yl]benzoic acid Ethyl ester, Ethyl 2-[3,6-Bis(ethylamino)-2,7-dimethyl-9H-xanthen-9-yl]benzoate

D5489-25MG D5489-100MG
TCIAD5489-25MGEA 221 EUR
TCIAD5489-25MG TCIAD5489-100MG
Dihydrorhodamine 6G
Dihydrorhodamine 6G
Formula: C₂₈H₃₂N₂O₃
Peso molecolare: 444,58 g/mol
Temperatura di stoccaggio: Refrigerata
CAS Number: 217176-83-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light red powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 6.0 to 6.6 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR