1,2-Dibromo-4-nitrobenzene ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
D5486-1G
D5486-5G
TCIAD5486-1GEA
39
EUR
TCIAD5486-1G
TCIAD5486-5G
1,2-Dibromo-4-nitrobenzene ≥98.0% (by GC)
1,2-Dibromo-4-nitrobenzene
Formula:
C₆H₃Br₂NO₂ Peso molecolare: 280,9 g/mol Punto di ebollizione: 296°C |
MDL Number:
MFCD00055373 CAS Number: 5411-50-7 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 55.0 to 59.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...