Stampa…

2,6-Dibromobenzoic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

D5485-1G D5485-5G
TCIAD5485-1GEA 61 EUR
TCIAD5485-1G TCIAD5485-5G
2,6-Dibromobenzoic acid ≥98.0% (by GC, titration analysis)
2,6-Dibromobenzoic acid
Formula: C₇H₄Br₂O₂
Punto di ebollizione: 335°C
MDL Number: MFCD00234254
CAS Number: 601-84-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 150.0 to 154.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR