DMOG ≥97.0% (by GC)
Fornitore: TCI
Synonyms:
N-[Methoxy(oxo)acetyl]glycine methyll ester, Dimethyloxaloylglycine, Methyl 2-((2-methoxy-2-oxoethyl)amino)-2-oxoacetate, DMOG
D5480-50MG
D5480-250MG
TCIAD5480-50MGEA
62
EUR
TCIAD5480-50MG
TCIAD5480-250MG
DMOG ≥97.0% (by GC)
DMOG
Formula:
C₆H₉NO₅ Peso molecolare: 175.14 g/mol Punto di ebollizione: 182°C |
MDL Number:
MFCD05865098 CAS Number: 89464-63-1 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 49.0 to 53.0 deg-C |
Acid Value | max. 10.0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...