Stampa…

β-Phenylpropiophenone ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: beta-Phenylpropiophenone, 1,3-Diphenyl-1-propanone

D5479-1G D5479-5G
TCIAD5479-1GEA 16 EUR
TCIAD5479-1G TCIAD5479-5G
β-Phenylpropiophenone ≥98.0% (by GC)
β-Phenylpropiophenone
Formula: C₁₅H₁₄O
Peso molecolare: 210,28 g/mol
Punto di ebollizione: 360°C
MDL Number: MFCD00039563
CAS Number: 1083-30-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 70.0 to 74.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR