3,5-Lutidine-N-oxide ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3,5-Lutidine N-Oxide, 3,5-Dimethylpyridine 1-oxide, 3,5-Dimethylpyridine N-Oxide
D5469-25G
D5469-5G
TCIAD5469-25GEA
357
EUR
TCIAD5469-25G
TCIAD5469-5G
3,5-Lutidine-N-oxide ≥98.0% (by GC)
3,5-Lutidine-N-oxide
Formula:
C₇H₉NO Peso molecolare: 123,1525 g/mol Punto di ebollizione: 168°C |
MDL Number:
MFCD00126839 CAS Number: 3718-65-8 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 104.0 to 108.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...