Stampa…
1,3-Dimethyl-1H-pyrazole-5-carboxylic acid ≥98.0% (by GC)
Fornitore: TCI EUROPE
Attenzione
D5468-25G D5468-5G
TCIAD5468-25GEA 180 EUR
TCIAD5468-25G TCIAD5468-5G
1,3-Dimethyl-1H-pyrazole-5-carboxylic acid ≥98.0% (by GC)
1,3-Dimethyl-1H-pyrazole-5-carboxylic acid
Formula: C₆H₈N₂O₂
Peso molecolare: 140,1400 g/mol
MDL Number: MFCD00159636
CAS Number: 5744-56-9
Visualizza i codici
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 96.0 %
Melting point 208.0 to 212.0 deg-C