Stampa…

Dihydromyricetin ≥97.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: (2R,3R)-3,5,7-Trihydroxy-2-(3,4,5-trihydroxyphenyl)-2,3-dihydrochromen-4-one, Ampelopsin, (2R,3R)-3,5,7-Trihydroxy-2-(3,4,5-trihydroxyphenyl)chroman-4-one

D5464-200MG D5464-1G
TCIAD5464-200MGEA 96 EUR
TCIAD5464-200MG TCIAD5464-1G
Dihydromyricetin ≥97.0% (by HPLC)
Dihydromyricetin
Formula: C₁₅H₁₂O₈
Peso molecolare: 320,25 g/mol
Punto di fusione: 248°C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00189451
CAS Number: 27200-12-0
Merck Index: 14,00579

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Specific rotation [a]20/D +15.5 to +17.0 deg (C=0.9, methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR