Dimethyl-2-aminoterephthalate ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
|
Synonyms:
Dimethyl aminoterephthalate, 2-Aminodimethyl terephthalate
Formula:
C₁₀H₁₁NO₄ Peso molecolare: 209.2 g/mol |
MDL Number:
MFCD00008427 CAS Number: 5372-81-6 EINECS: 226-364-2 UN: 3077 ADR: 9 III |
Risultati del test di specificazione
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 133.0 to 137.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...