Stampa…

4,4'-Dimethylbenzhydrol ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Bis(4-methylphenyl)methanol, 4,4'-Dimethylbenzhydryl alcohol, Di-p-tolylmethanol, 4,4'-Dimethyl-α-phenylbenzyl alcohol, Bis(4-methylphenyl)carbinol, 4,4'-Dimethylphenyl phenyl carbinol, 4,4'-Dimethyldiphenylmethanol

D5460-1G D5460-5G
TCIAD5460-1GEA 65 EUR
TCIAD5460-1G TCIAD5460-5G
4,4'-Dimethylbenzhydrol ≥98.0% (by GC)
4,4'-Dimethylbenzhydrol
Formula: C₁₅H₁₆O
Peso molecolare: 212,29 g/mol
MDL Number: MFCD00017216
CAS Number: 885-77-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5768452a85e9962920656368ba890f6bc65d1630&vid=5768452a85e9962920656368ba890f6bc65d1630&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 69.0 to 73.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR