Stampa…

4,6-Di(tert-butyl)benzene-1,3-diol ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4,6-Di-tert-butylresorcinol, 4,6-Di-tert-butylbenzene-1,3-diol

D5459-25G D5459-5G
TCIAD5459-25GEA 200 EUR
TCIAD5459-25G TCIAD5459-5G
4,6-Di(tert-butyl)benzene-1,3-diol ≥98.0% (by GC)
4,6-Di(tert-butyl)benzene-1,3-diol
Formula: C₁₄H₂₂O₂
Peso molecolare: 222,33 g/mol
MDL Number: MFCD00044513
CAS Number: 5374-06-1
EINECS: 226-366-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 121.0 to 125.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR