Stampa…

Diphenyl Phenylphosphonate ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Phenylphosphonic acid Diphenyl ester

D5454-200MG D5454-1G
TCIAD5454-200MGEA 76 EUR
TCIAD5454-200MG TCIAD5454-1G
Diphenyl Phenylphosphonate ≥98.0% (by GC)
Diphenyl Phenylphosphonate
Formula: C₁₈H₁₅O₃P
Peso molecolare: 310,29 g/mol
Punto di ebollizione: 165°C
Punto di fusione: 75°C
MDL Number: MFCD00216888
CAS Number: 3049-24-9
EINECS: 221-261-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d8c92662854fb854bc083eff6c40d3d93d5272c&vid=4d8c92662854fb854bc083eff6c40d3d93d5272c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 72.0 to 76.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR