Stampa…

2,7-Dibromo-9-phenyl-9H-fluoren-9-ol ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 2,7-Dibromo-9-hydroxy-9-phenyl-9H-fluorene

D5448-5G D5448-25G
TCIAD5448-5GEA 78 EUR
TCIAD5448-5G TCIAD5448-25G
2,7-Dibromo-9-phenyl-9H-fluoren-9-ol ≥98.0% (by GC)
2,7-Dibromo-9-phenyl-9H-fluoren-9-ol
Formula: C₁₉H₁₂Br₂O
Peso molecolare: 416,11 g/mol
MDL Number: MFCD09953789
CAS Number: 132717-37-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=61d2217146f2dddaf81d79e50047edcb7b95dc28&vid=61d2217146f2dddaf81d79e50047edcb7b95dc28&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 164.0 to 168.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR