Stampa…

6,6''-Dibromo-2,2':6',2''-terpyridine ≥94.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

D5432-200MG D5432-1G
TCIAD5432-200MGEA 100 EUR
TCIAD5432-200MG TCIAD5432-1G
6,6''-Dibromo-2,2':6',2''-terpyridine ≥94.0% (by GC)
6,6''-Dibromo-2,2':6',2''-terpyridine
Formula: C₁₅H₉Br₂N₃
Peso molecolare: 391,07 g/mol
MDL Number: MFCD01863557
CAS Number: 100366-66-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR