Stampa…

3,5-Dinitrophenylboronic acid pinacol ester ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 1,3-Dinitro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 2-(3,5-Dinitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,5-Dinitrophenylboronic acid Pinacol ester

D5431-200MG D5431-1G
TCIAD5431-200MGEA 95 EUR
TCIAD5431-200MG TCIAD5431-1G
3,5-Dinitrophenylboronic acid pinacol ester ≥98.0% (by GC)
3,5-Dinitrophenylboronic acid pinacol ester
Formula: C₁₂H₁₅BN₂O₆
Peso molecolare: 294,07 g/mol
MDL Number: MFCD11504965
CAS Number: 428820-95-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7040112e25d928a1d421fe5db66cab5de41d3aa1&vid=7040112e25d928a1d421fe5db66cab5de41d3aa1&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 96.0 %
Melting point 147.0 to 151.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR