10,15-Dihydro-5H-diindolo[3,2-a:3',2'-c]carbazole ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
10,15-Dihydro-5H-5,10,15-triazadiindeno[1,2-a:1',2'-c]fluorene
D5428-200MG
D5428-1G
TCIAD5428-200MGEA
65
EUR
TCIAD5428-200MG
TCIAD5428-1G
10,15-Dihydro-5H-diindolo[3,2-a:3',2'-c]carbazole ≥98.0% (by HPLC)
10,15-Dihydro-5H-diindolo[3,2-a:3',2'-c]carbazole
Formula:
C₂₄H₁₅N₃ Peso molecolare: 345,41 g/mol Punto di fusione: 350°C |
MDL Number:
MFCD21609440 CAS Number: 109005-10-9 |
Risultati del test di specificazione
Appearance | White to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...