Stampa…

3,3-Dimethyl-3,4-dihydro-2H-thieno[3,4-b][1,4]dioxepine ≥95.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 3,4-(2,2'-Dimethylpropylenedioxy)thiophene

D5423-200MG D5423-1G
TCIAD5423-200MGEA 61 EUR
TCIAD5423-200MG TCIAD5423-1G
3,3-Dimethyl-3,4-dihydro-2H-thieno[3,4-b][1,4]dioxepine ≥95.0% (by GC)
3,3-Dimethyl-3,4-dihydro-2H-thieno[3,4-b][1,4]dioxepine
Formula: C₉H₁₂O₂S
Peso molecolare: 184,25 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD08705268
CAS Number: 255901-50-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d5a7c2f9893918ce0a0c4353bb67afc100fe535&vid=8d5a7c2f9893918ce0a0c4353bb67afc100fe535&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green powder to lump
Purity (GC) min. 95.0 %
Melting point 42.0 to 46.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR