Stampa…

Diazoxide ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-Methyl-7-chloro-1,2,4-benzothiadiazine 1,1-dioxide, 7-Chloro-3-methyl-2H-1,2,4-benzothiadiazine 1,1-Dioxide

D5402-250MG D5402-1G
TCIAD5402-250MGEA 64 EUR
TCIAD5402-250MG TCIAD5402-1G
Diazoxide ≥98.0% (by HPLC, titration analysis)
Diazoxide
Formula: C₈H₇ClN₂O₂S
Peso molecolare: 230,666 g/mol
MDL Number: MFCD00078578
CAS Number: 364-98-7
EINECS: 206-668-1
Merck Index: 14,03004
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b87d2d035ea29b779fc57cbb5114d16201cf121&vid=7b87d2d035ea29b779fc57cbb5114d16201cf121&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR