Stampa…

3-Diethylamino-1-propyne ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: N,N-Diethylpropargylamine

D5393-5ML D5393-25ML
TCIAD5393-5MLEA 44 EUR
TCIAD5393-5ML TCIAD5393-25ML
3-Diethylamino-1-propyne ≥98.0% (by GC, titration analysis)
3-Diethylamino-1-propyne
Formula: C₇H₁₃N
Peso molecolare: 111,19 g/mol
Punto di ebollizione: 120°C
Flash Pt: 20°C
MDL Number: MFCD00039881
CAS Number: 4079-68-9
EINECS: 223-804-5
UN: 2733
ADR: 3,II

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR