Stampa…

3-(Dimethylamino)-1-phenyl-2-propen-1-one ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE

D5384-1G D5384-5G
TCIAD5384-1GEA 52 EUR
TCIAD5384-1G TCIAD5384-5G
3-(Dimethylamino)-1-phenyl-2-propen-1-one ≥98.0% (by GC, titration analysis)
3-(Dimethylamino)-1-phenyl-2-propen-1-one
Formula: C₁₁H₁₃NO
Peso molecolare: 175,23 g/mol
MDL Number: MFCD00121193
CAS Number: 1201-93-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b2693dd11da9438f0d3db1df07b419622113bbf&vid=0b2693dd11da9438f0d3db1df07b419622113bbf&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 90.0 to 94.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR