1-Decyl-3-methylimidazoliumtetrafluoroborate ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI
|
Attenzione
|
Synonyms:
1-Decyl-3-methylimidazolium tetrafluoroborate
Formula:
C₁₄H₂₇BF₄N₂ Peso molecolare: 310.19 g/mol Punto di fusione: –4°C |
MDL Number:
MFCD09038853 CAS Number: 244193-56-4 |
Risultati del test di specificazione
Appearance | Light yellow to Brown clear liquid |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...