Dehydrocostus Lactone ≥98.0% (by GC)
Fornitore: TCI
Synonyms:
(3aS,6aR,9aR,9bS)-3,6,9-Trimethylenedecahydroazuleno[4,5-b]furan-2(3H)-one, (3aS,6aR,9aR,9bS)-3,6,9-Trimethylenedecahydroazuleno[4,5-b]furan-2(9bH)-one
Formula:
C₁₅H₁₈O₂ Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00210277 CAS Number: 477-43-0 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 57.0 to 61.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...