4,4'-Dibromo-4''-methoxytriphenylamine ≥98.0% (by total nitrogen basis)
Fornitore: TCI
Formula:
C₁₉H₁₅Br₂NO |
MDL Number:
MFCD31699798 CAS Number: 100308-69-8 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 73.0 to 77.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...