1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
EDC Methiodide, 3-(((Ethylimino)methylene)amino)-N,N,N-trimethylpropan-1-aminium iodide, 1-[3-(Dimethylamino)propyl]-3-ethylcarbodiimide Methiodide
D5334-5G
D5334-25G
TCIAD5334-5GEA
50
EUR
TCIAD5334-5G
TCIAD5334-25G
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide ≥98.0% (by titrimetric analysis)
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide
Formula:
CH₃CH₂N=C=N(CH₂)₃N(CH₃)₃I Peso molecolare: 297,17 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00011833 CAS Number: 22572-40-3 EINECS: 245-095-1 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 95.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 95.0 to 99.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...