Stampa…
4,5-Dichloro-2-phenyl-3(2H)-pyridazinone ≥98.0% (by GC)
Fornitore: TCI EUROPE
Attenzione
Synonyms: 4,5-Dichloro-2-phenylpyridazin-3(2H)-one
D5325-1G D5325-5G
TCIAD5325-1GEA 21 EUR
TCIAD5325-1G TCIAD5325-5G
4,5-Dichloro-2-phenyl-3(2H)-pyridazinone ≥98.0% (by GC)
4,5-Dichloro-2-phenyl-3(2H)-pyridazinone
Formula: C₁₀H₆Cl₂N₂O
Peso molecolare: 241,07 g/mol
MDL Number: MFCD00006470
CAS Number: 1698-53-9
EINECS: 216-917-6
Visualizza i codici
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 163.0 to 167.0 deg-C