2,4-Dibromo-6-nitrophenol ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1,3-Dibromo-4-hydroxy-5-nitrobenzene
D5323-1G
D5323-5G
TCIAD5323-1GEA
79
EUR
TCIAD5323-1G
TCIAD5323-5G
2,4-Dibromo-6-nitrophenol ≥98.0% (by GC)
2,4-Dibromo-6-nitrophenol
Formula:
C₆H₃Br₂NO₃ Peso molecolare: 296,9 g/mol |
MDL Number:
MFCD00024232 CAS Number: 15969-09-2 |
Risultati del test di specificazione
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 116.0 to 120.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...