1,4-Dibromo-2,5-bis[2-(trimethylsilyl)ethynyl]benzene ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
[(2,5-Dibromo-1,4-phenylene)bis(ethyne-2,1-diyl)]bis(trimethylsilane), [2-[2,5-Dibromo-4-[2-(trimethylsilyl)ethynyl]phenyl]ethynyl]trimethylsilane
D5316-1G
D5316-5G
TCIAD5316-1GEA
116
EUR
TCIAD5316-1G
TCIAD5316-5G
1,4-Dibromo-2,5-bis[2-(trimethylsilyl)ethynyl]benzene ≥98.0% (by GC)
1,4-Dibromo-2,5-bis[2-(trimethylsilyl)ethynyl]benzene
Formula:
C₁₆H₂₀Br₂Si₂ Peso molecolare: 428,31 g/mol |
MDL Number:
MFCD11521283 CAS Number: 478190-79-3 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 119.0 to 123.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...