Stampa…

1,4-Diethynyl-2,5-dimethoxybenzene ≥98.0% (by GC)

Fornitore: TCI EUROPE

D5315-1G D5315-200MG
TCIAD5315-1GEA 198 EUR
TCIAD5315-1G TCIAD5315-200MG
1,4-Diethynyl-2,5-dimethoxybenzene ≥98.0% (by GC)
1,4-Diethynyl-2,5-dimethoxybenzene
Formula: C₁₂H₁₀O₂
Peso molecolare: 186,21 g/mol
Punto di fusione: 166°C
MDL Number: MFCD00846857
CAS Number: 74029-40-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR