Stampa…

1,4-Diethynyl-2,5-dimethylbenzene ≥98.0% (by GC)

Fornitore: TCI EUROPE

D5314-200MG D5314-1G
TCIAD5314-200MGEA 159 EUR
TCIAD5314-200MG TCIAD5314-1G
1,4-Diethynyl-2,5-dimethylbenzene ≥98.0% (by GC)
1,4-Diethynyl-2,5-dimethylbenzene
Formula: C₁₂H₁₀
Peso molecolare: 154,21 g/mol
CAS Number: 75867-45-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 110.0 to 114.0 deg-C
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR