(E)-4-(Dimethylamino)chalcone ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
Synonyms:
2-(4-Dimethylaminobenzal)acetophenone, 2-(4-Dimethylaminobenzylidene)acetophenone, (E)-3-[4-(Dimethylamino)phenyl]-1-phenylprop-2-en-1-one
D5312-200MG
D5312-1G
TCIAD5312-200MGEA
87
EUR
TCIAD5312-200MG
TCIAD5312-1G
(E)-4-(Dimethylamino)chalcone ≥98.0% (by GC, titration analysis)
(E)-4-(Dimethylamino)chalcone
Formula:
C₁₇H₁₇NO Peso molecolare: 251,33 g/mol |
CAS Number:
22965-98-6 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 112.0 to 116.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...