Stampa…

Dimethyl-3-nitrophthalate ≥99.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Dimethyl 3-nitrophthalate

D5310-5G D5310-25G
TCIAD5310-5GEA 96 EUR
TCIAD5310-5G TCIAD5310-25G
Dimethyl-3-nitrophthalate ≥99.0% (by GC)
Dimethyl-3-nitrophthalate
Formula: C₁₀H₉NO₆
Peso molecolare: 239,19 g/mol
MDL Number: MFCD00017184
CAS Number: 13365-26-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=844fb364207344ad7760e6437b042e465f3b24c3&vid=844fb364207344ad7760e6437b042e465f3b24c3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Melting point 66.0 to 70.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR